missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: Super I, Basic pH, MP EcoChrom™, MP Biomedicals™

Catalog No. ICN04577
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN04577 50 kg
ICN04571 1 kg
ICN04574 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN04577 Supplier MP Biomedicals Inc Supplier No. 0204577
Only null left

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This Alumina is produced at the highest activity available in the market.

  • MP EcoChrom™ Alumina - Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
  • MP Alumina B - Super I show absorption capacity approximately twice as high as an Alumin B, Activity I.
  • The initial activity is kept in a very narrow range and their deactivation behaviour is extremely constant.
  • The product is available as a neutral version, MP Alumina N - Super I, and an acidic version, MP Alumina A - Super I.
  • MP EcoChro Alumina B - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides, environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 50 kg
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I
EINECSNumber 215-691-6
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.