missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: I, MP EcoChrom™, MP Biomedicals™

Catalog No. p-4650839
Change view
Click to view available options
Quantity:
1 kg
50 kg
500 g
Catalog No. Quantity
ICN02069 500 g
ICN02072 1 kg
ICN02078 50 kg
3 options

Catalog No. ICN02069

Supplier: MP Biomedicals Inc 0202069

Only null left

MP EcoChrom™ Alumina B, Activity I are good all-round adsorbents for laboratory use as well as for industrial use. They can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, (e.g. glycol or glycerol can be used).

  • MP EcoChrom™ Alumina B, Activity I is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc.
  • It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: I, Activity acc. To Brockmann: 24 l/mm
EINECSNumber 215-691-6
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.