missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, 32-63μM, Active, MP Biomedicals™

Catalog No. ICN02040
Catalog No. ICN02040 Supplier MP Biomedicals Inc Supplier No. 0202040

For Use in Column Chromatography and Adsorptive Filtration

Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories

Specifications

Quantity 500 g
Content And Storage Store at Room Temperature.
Type Alumina B
Physical Form Powder
MDL Number MFCD00003424
Density 3.94g/cm3 at 20°C (Lit.)
CAS 1344-28-1