missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina, Activity: II-III, MP EcoChrom™, MP Biomedicals™
Use MP EcoChrom™ Alumina, Activity II-III as an adsorbent, or in chromatography and thin layer chromatography (TLC).
$638.89 - $638.89
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| IUPAC Name | dialuminum;oxygen(2-) |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
- MP EcoChrom™ Alumina, Activity II-III is an economical adsorbents of medium activity.
- It is often used where activated charcol can not be used due to its organic nature, or, where the cation exchange properties of basic alumina are desired.
- It is suitable for the development of separation methods and for technical separations where a custom-made adsorbents for special separation is not available.
- An oxide of aluminum, it occurs in nature as various minerals such as bauxite, corundum, etc.
- It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| dialuminum;oxygen(2-) |
Specifications
| Pink to White | |
| Free-flowing Powder | |
| Al2O3 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| 101.96 | |
| Alumina, Activity: II-III, Activity acc to Brockmann: 17/mm |
| 1344-28-1 | |
| 5 kg | |
| MFCD00003424 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminum;oxygen(2-) | |
| 9989226 | |
| 3.94 g/cm3 (at 20°C) |
Safety and Handling
EINECSNumber : 215-691-6
Recommended Storage : Store at room temperature.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.