missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina A, Activity: I, MP EcoChrom™, MP Biomedicals™
MP EcoChrom™ Alumina A catalyses a variety of reactions that are useful industrially.
$226.13 - $226.13
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
- Alumina A is widely used to remove water from gas streams.
- Also used for its hardness and strength.
- It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
- Many types of sandpaper use aluminium oxide crystals.
- In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations, as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments.
- Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides.
- Acidic alumina, is the weakest and least often used type.
- Alumina is useful for separation of acid pigments and strong acids.
- It can cause polymerization, condensation and dehydration reactions.
- Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
- It is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| dialuminium(3+) trioxidandiide |
Specifications
| Pink to White | |
| Free-flowing Powder | |
| Al2O3 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| 101.96 | |
| Alumina A, Activity: I, Activity acc. To Brockmann: 23 l/mm |
| 1344-28-1 | |
| 500 g | |
| MFCD00003424 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| 9989226 | |
| 3.94 g/cm3 (at 20°C) |
Safety and Handling
Recommended Storage : Store at room temperature.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.