missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alum Solution, 10% (w/v), for Dissolved Oxygen, LabChem™
Specifications
Specifications
| Color | Clear, colorless |
| Format | Liquid |
| Packaging | Poly Bottle |
| Quantity | 1 L |
| Storage Requirements | Room Temperature |
| Type | Alum Solution |