missing translation for 'onlineSavingsMsg'
Learn More

Alum Solution, 10% (w/v), for Dissolved Oxygen, LabChem™

Catalog No. LC107401
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC107401 500 mL
LC107402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC107401 Supplier LabChem Supplier No. LC107401
Only null left

Specifications

Color Clear, colorless
Format Liquid
Packaging Poly Bottle
Quantity 500 mL
Storage Requirements Room Temperature
Type Alum Solution