missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Altrenogest 98.0+%, TCI America™
$106.25 - $359.76
Chemical Identifiers
| CAS | 850-52-2 |
|---|---|
| Molecular Formula | C21H26O2 |
| Molecular Weight (g/mol) | 310.437 |
| MDL Number | MFCD00867855 |
| InChI Key | VWAUPFMBXBWEQY-MXEMCNAFSA-N |
| Synonym | 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one, Allyltrenbolone |
| PubChem CID | 129317581 |
| IUPAC Name | (8S,13S,14R,17S)-17-hydroxy-13-methyl-17-prop-2-enyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
| SMILES | CC12C=CC3=C4CCC(=O)C=C4CCC3C1CCC2(CC=C)O |
Chemical Identifiers
| 850-52-2 | |
| 310.437 | |
| VWAUPFMBXBWEQY-MXEMCNAFSA-N | |
| 129317581 | |
| CC12C=CC3=C4CCC(=O)C=C4CCC3C1CCC2(CC=C)O |
| C21H26O2 | |
| MFCD00867855 | |
| 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one, Allyltrenbolone | |
| (8S,13S,14R,17S)-17-hydroxy-13-methyl-17-prop-2-enyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
Specifications
| 850-52-2 | |
| Yellow | |
| MFCD00867855 | |
| 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one, Allyltrenbolone | |
| CC12C=CC3=C4CCC(=O)C=C4CCC3C1CCC2(CC=C)O | |
| 310.437 | |
| 310.44 | |
| Crystalline Powder |
| 118°C | |
| C21H26O2 | |
| 200 mg | |
| VWAUPFMBXBWEQY-MXEMCNAFSA-N | |
| (8S,13S,14R,17S)-17-hydroxy-13-methyl-17-prop-2-enyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one | |
| 129317581 | |
| ≥98.0% (HPLC) | |
| Altrenogest |
Safety and Handling
RTECSNumber : KG7745000
TSCA : No
Recommended Storage : Refrigerator