missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers

Catalog No. 13762397
Change view
Click to view available options
Type:
Standard Plug (Factory Installed)
Twist Lock (Factory Installed)
Twist Lock (Field Installed)
Plug Type:
US NEMA 6-15 P
US NEMA 6-20 P
US NEMA L6-15 P
Amperage:
15 A
20 A
Voltage:
230 V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Plug Type Amperage Voltage
13762397 Standard Plug (Factory Installed) US NEMA 6-15 P 15 A 230 V
430267 Twist Lock (Factory Installed) US NEMA L6-15 P 15 A 230 V
13762394 Twist Lock (Field Installed) US NEMA 6-20 P 20 A 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13762397 Supplier Thermo Scientific™ Supplier No. TL230V15A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers.

Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers. Options include an array of standard and special plugs for North America and international markets.

Specifications

Type Standard Plug (Factory Installed)
For Use With (Equipment) Thermo Scientific™ TDE and TSX Series,Thermo Scientific™ TSU Series, HERAfreeze™ HFU-T, Revco™ UxF
Amperage 15 A
Plug Type US NEMA 6-15 P
Voltage 230 V