missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alsever's Solution

Catalog No. AAJ67805AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67805AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67805AP Supplier Thermo Scientific Chemicals Supplier No. J67805AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Alsevers Solution
Color Colorless
Physical Form Liquid
Quantity 500 mL

RUO – Research Use Only