Learn More
Ansell™ AlphaTec™ PVC Coat Apron
Catalog No. 19010245
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19010245 X-Large
19010242 Small
19010243 Medium
19010244 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19010245 Supplier Ansell™ Supplier No. CAXL8
Only null left
Ansell™ AlphaTec™ PVC Coat Apron keeps workers clean and protected wherever oil, grease and chemicals are present
  • No exposed elastic or metal parts to cause static or corrosion
  • Protection when splashes from acids and caustic bases are a concern
  • Virgin vinyl construction contains no reprocessed materials
  • Flexible
  • Resistant to punctures, abrasion, chemicals, fats, oils and greases
  • Sleeves come with elasticized cuffs for added protection
  • PVC (vinyl) grommets are permanently sealed to the aprons for durability and allow for size adjustment without tear-out
  • Six sizes with adjustable ties ensure optimal fit for a wide range of users
Specifications
Size X-Large
Material PVC
Color Blue
Disposable Reusable
Closure Type Ties
Sterility Non-sterile
Cuff Style Elastic
Latex Free No
Thickness 8 mil
For Use With (Application) Packing of filets and ground meat; Canning; Bottling; Storing and transportation; Equipment maintenance and repair; Plant disinfection and sanitization; Transferring liquids
Seam Type Dielectrically Sealed
Show More Show Less