missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ PVC Coat Apron
Description
- No exposed elastic or metal parts to cause static or corrosion
- Protection when splashes from acids and caustic bases are a concern
- Virgin vinyl construction contains no reprocessed materials
- Flexible
- Resistant to punctures, abrasion, chemicals, fats, oils and greases
- Sleeves come with elasticized cuffs for added protection
- PVC (vinyl) grommets are permanently sealed to the aprons for durability and allow for size adjustment without tear-out
- Six sizes with adjustable ties ensure optimal fit for a wide range of users
Specifications
Specifications
| Size | Large |
| Material | PVC |
| Color | Blue |
| Disposable | Reusable |
| Closure Type | Ties |
| Sterility | Non-sterile |
| Cuff Style | Elastic |
| Latex Free | No |
| Thickness | 8 mil |
| For Use With (Application) | Packing of filets and ground meat; Canning; Bottling; Storing and transportation; Equipment maintenance and repair; Plant disinfection and sanitization; Transferring liquids |
| Show More |