missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood and Socks
Description
- Protection: Barrier to numerous organic and inorganic chemicals and biological hazards; >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Comfort: Multi-layer material which is lightweight, yet strong and durable
- Highly visible: Bright orange color for improved worker safety
- Design: Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece hood, integrated socks with boot overflap
- Applications: Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, mining, agriculture, HAZMAT emergency reponse (Level B), first response and fire service
- Sizes: Available in Small through 5X-Large
- Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
Specifications
Specifications
| Size | X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Orange |
| Waist Style | Elastic |
| Ankle Style | Attached Socks, Boot Overflaps |
| Cuff Style | Elastic, Knit |
| Neck Style | Hooded |
| Hood Type | 2-panel |
| Closure Type | Double Zipper |
| Seam Type | Welded, Taped |
| Show More |