missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood

Provide an extremely effective barrier against both chemical and biological hazards. Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood are durable and multi-layered, and are some of the most comfortable chemical protective materials available.

Supplier:  Ansell™ YE30W9211104

 View more versions of this product

Catalog No. 17-999-926


Only null left
Explore available promotions

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Description

Description

  • Protection:  Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible:  Bright yellow for improved worker safety
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs, ultrasonically welded seams, finger loops, 2 piece respirator fit hood
  • Applications:  Chemicals, oil and petrochemicals, pharmaceutical, food indsutry, sewage purification installations, industrial and tank cleaning, mining
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Specifications

Large
Yellow
Elastic
Hooded
Double Zipper
Liquid and spray tight chemical and biological protection
Multi-layer Barrier Laminate Fabric
Elastic
Double, Elastic
3-panel, Respirator Fit
Welded
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions