missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Collar

Catalog No. 17999900
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999900 Small
17999901 Medium
17999902 Large
17999903 X-Large
17999904 2X-Large
17999905 3X-Large
17999907 5X-Large
17999906 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17999900 Supplier Ansell™ Supplier No. YY23B9210302
This product contains natural rubber latex which may cause allergic reactions.

Provide an excellent barrier to harmful chemicals and blood borne pathogens. Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Collar are lightweight and relatively strong and durable. These coveralls are designed for applications such as environmental clean-up, sewage purification installations, and composites manufacturing.

  • Protection:  A barrier to numerous inorganic liquid chemicals including acids and bases; achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 & ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens
  • Comfort:  Lightweight yet strong and durable
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, bound seams, finger loops, collar
  • Applications:  Environmental cleanup, sewage purification installations, composites manufacturing
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671

Specifications

Size Small
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Thumb Loop
Neck Style Collared
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Bound
Protection Class Particulate and light liquid splash protection
Latex Free No
Show More Show Less