missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 2000 Standard Bound Coveralls with Hood, Model 111

Catalog No. 17999876
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999876 Small
17999877 Medium
17999878 Large
17999879 X-Large
17999880 2X-Large
17999883 5X-Large
17999882 4X-Large
17999881 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17999876 Supplier Ansell Supplier No. WH20B9211102

Coveralls with breathable microporous laminate technology are ideal for a wide range of industrial applications. Previously known as MICROCHEM™ by AlphaTec 2000 Standard.

  • Liquid penetration resistance and barrier to fine particulates
  • Compliant with ASTM F1761 and EN 14126:2003 resistance to penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static properties, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic wrists and ankles
  • 2-way front zipper with storm flap
  • Bound seams, finger loops
  • 3-piece hood
  • Applications include pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigations, veterinary services, food
  • Available in small through 5X-large sizes
  • EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Particle, Liquid, Chemicals, Anti-static
Certifications/Compliance ASTM F1761, AATCC76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 14126:2003, EN1149-5 , DIN 32781, EN 1073-2, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
Latex Free Yes
Model WH20-B-92-111
For Use With (Application) Life Sciences, Automotive, Metal Fabrication, Food Processing
Show More Show Less