missing translation for 'onlineSavingsMsg'
Learn More

Ansell Alphatec™ 1500 Stitched Model 106, Coveralls with Hood and Boots

Catalog No. 17999852
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999852 Small
17999853 Medium
17999854 Large
17999855 X-Large
17999856 2X-Large
17999857 3X-Large
17999859 5X-Large
17999858 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17999852 Supplier Ansell Supplier No. WH15S9210602
This product contains natural rubber latex which may cause allergic reactions.

Particle-filtering SMS protective suit with attached hood and boots offers wearers light chemical resistance and breathable comfort. Previously known as MICROCHEM™.

  • Proven to filter 100% of particles over 3 microns in size
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free, critical in spray painting applications
  • Optimized body fit
  • Elastic wrists and ankles with attached boots, non-slip soles
  • 2-way front zipper with storm flap, stitched seams
  • 3 piece respirator fit hood
  • Ideal for asbestos related work, handling powders, general maintenance, construction
  • Available in small through 5X-large sizes
  • ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid
Collar Type None
Certifications/Compliance (Europe) CE Category III (Type 5, Type 6, EN 1073-2)
Latex Free No
Model WH15-S-92-106
For Use With (Application) Inspection, Asbestos Related Work, General Maintenance, Powder Handling
Height (English) 5 ft. to 5 ft. 7 in.
Quantity 25/Cs.
Packaging Individually Packaged
Show More Show Less