Learn More
Ansell Alphatec™ 1500 Stitched Model 101, Coveralls with Hood
Particle-filtering SMS protective suit with hood offers wearers light chemical resistance and breathable comfort. Previously known as MICROCHEM™.
Supplier:  Ansell WH15S9210104
 View more versions of this product
Catalog No. 17-999-846
Only null left
Explore available promotions
Alert:
This product contains natural rubber latex which may cause allergic reactions.
Description
Description
  • Proven to filter 100% of particles over 3 microns in size
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free, critical in spray painting applications
  • Optimized body fit
  • Elastic wrists and ankles without attached boots
  • 2-way front zipper with storm flap, stitched seams
  • 3 piece elasticated hood
  • Ideal for asbestos related work, handling powders, general maintenance, construction
  • Available in small through 5X-large sizes
  • ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 8
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
3-panel, Respirator Fit
Stitched
Non-toxic Particulates, Liquid
CE Category III (Type 5, Type 6, EN 1073-2)
WH15-S-92-101
5 ft. 5 in. to 5 ft. 10 in.
Individually Packaged
Large
SMS Fabric
Single-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
None
No
Inspection, Asbestos Related Work, General Maintenance, Powder Handling, Construction
25/Cs.
Product Suggestions
Product Suggestions