missing translation for 'onlineSavingsMsg'
Learn More

Ansell Alphatec™ 1500 Stitched Model 100, Coveralls with Collar

Catalog No. p-7070768
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999837 Medium
17999839 X-Large
17999838 Large
17999836 Small
17999840 2X-Large
17999841 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17999837 Supplier Ansell Supplier No. WH15S9210003
Only null left
This product contains natural rubber latex which may cause allergic reactions.

Particle-filtering SMS protective suit with collar offers wearers light chemical resistance and breathable comfort. Previously known as MICROCHEM™.

  • Proven to filter 100% of particles over 3 microns in size
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free, critical in spray painting applications
  • Optimized body fit
  • Elastic wrists and ankles without attached boots
  • 2-way front zipper with storm flap, stitched seams
  • Collar only, no hood
  • Ideal for asbestos related work, handling powders, general maintenance, construction
  • Available in small through 5X-large sizes
  • ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid
Collar Type Collared
Certifications/Compliance (Europe) CE Category III (Type 5, Type 6, EN 1073-2)
Latex Free No
Model WH15-S-92-100
For Use With (Application) Inspection, Asbestos Related Work, General Maintenance, Powder Handling, Construction
Height (English) 5 ft. 2 in. to 5 ft. 7 in.
Quantity 25/Cs.
Packaging Individually Packaged
Show More Show Less