missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Hood

Designed for protection from particulates and light, non-flammable liquid splashes. Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Hood are made of highly breathable, flame retardant and anti-static SMMS. They are perfect for work in utilities, industrial cleaning, general maintenance, and the petrochemical industry.

Supplier:  Ansell™ NR17S9211106

 View more versions of this product

Catalog No. 18-000-000

Only null left
Explore available promotions

Alert:

This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.


Description

Description

  • Protection:  Flame retardant and anti-static SMMS nonwoven provides a good barrier to particulates and low hazard liquid splashes
  • Comfort:  Air and water vapor permeable ("breathable") to help reduce the risk of heat stress
  • Optimized body fit:  Ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above)
  • Anti-static:  Tested according to EN1149-5
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, finger loops, red stitching for ease of identification, serged seams, 3 piece respirator fit hood
  • Applications:  Petrochemical industry, industrial cleaning, utilities, general maintenance
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 6
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 10

Specifications

Specifications

2X-Large
Navy, Red
Elastic
Hooded
2-way Front Zipper with Storm Flap
Particulate and light liquid splash protection
FR SMMS
Elastic
Elastic, Finger Loops
3-panel, Respirator Fit
Serged
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions