missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Collar

Catalog No. 17999989
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999989 Medium
17999990 Large
17999991 X-Large
17999992 2X-Large
17999993 3X-Large
17999994 4X-Large
17999995 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.
Catalog No. 17999989 Supplier Ansell™ Supplier No. NR17S9210303
This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.

Designed for protection from particulates and light, non-flammable liquid splashes. Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Collar are made of highly breathable, flame retardant and anti-static SMMS. They are perfect for work in utilities, industrial cleaning, general maintenance, and the petrochemical industry.

  • Protection:  Flame retardant and anti-static SMMS nonwoven provides a good barrier to particulates and low hazard liquid splashes
  • Comfort:  Air and water vapor permeable ("breathable") to help reduce the risk of heat stress
  • Optimized body fit:  Ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above)
  • Anti-static:  Tested according to EN1149-5
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, finger loops, red stitching for ease of identification, serged seams, collar
  • Applications:  Petrochemical industry, industrial cleaning, utilities, general maintenance
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 6
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 6

Specifications

Size Medium
Material FR SMMS
Color Navy, Red
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Collared
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged
Protection Class Particulate and light liquid splash protection
Latex Free No
Show More Show Less