missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ Medium Duty Splash Protection Vinyl Apron

Catalog No. 19010251
Change view
Click to view available options
Dimensions (L x W):
33 x 44 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19010251 33 x 44 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19010251 Supplier Ansell™ Supplier No. ECB45

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ansell™ AlphaTec™ Medium Duty Splash Protection Vinyl Apron keeps workers keep clean and protected from harmful contact wherever oil, grease and chemicals are present

  • PVC (Vinyl) aprons traditionally offer excellent protection where splashes from acids and caustic bases are a concern
  • Die-cut PVC construction features superior flexibility yet excellent resistance to abrasion and puncture
  • Integral ties provide an economical solution for adjustable fit
  • Applications:
    • Packing of fillets and ground meat
    • Canning
    • Bottling
    • Storing and transportation
    • Equiptment maintenance and repair
    • Plant disinfection and sanitization
    • Transferring liquids

Specifications

Type Apron
Material PVC
Color Blue
Closure Type Adjustable Ties
Sterility Non-sterile
Construction Die cut PVC
Latex Free No
Thickness 6 mil
Dimensions (L x W) 33 x 44 in.