missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 62-401 Latex Gloves, Cotton Lining

Catalog No. 19042380
Change view
Click to view available options
Size:
11
7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19042380 7
19042384 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19042380 Supplier Ansell Supplier No. M9999818
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Ansell AlphaTec™ Glove with Liquid-Proof Coating is comfortable for intensive use.

  • Heavy duty glove
  • Wrinkle finish for excellent wet and dry grip
  • Ergonomically styled fit providing maximum comfort
  • Cotton liner provide excellent thermal insulation (hot or cold)
  • Compliant with FDA food handling requirements

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009; Category lll, EN 407:02XXXX, EN 407:X2XXXX, EN 511:011
Color Blue
Latex Free No
Material Latex
Size 7
Thickness (English) 53 mil
Thickness (Metric) 1.35 mm
EN388 Standard EN 388:3131B; EN 388:3231
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Crinkle Finish
Liner Material Cotton
Lining Lined
Model 62401070
Length (English) 13 in.
Length (Metric) 32 cm
Show More Show Less