missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves

Catalog No. 19010377
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
19010377 4X-Large
19010371 Small
19010372 Medium
19010373 Large
19010374 X-Large
19010375 2X-Large
19010376 3X-Large
19010378 5X-Large
8 options

Catalog No. 19010377

Supplier: Ansell™ GR40T9221508

Only null left

Ansell™ AlphaTec™ Chemical Protective Apron with Sleeves for exceptional chemical protection

  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted inner cuff
  • Welded and taped seams
  • Applications:
    • Contaminated environments
    • Chemical handling
    • Filling, blending and charging of raw materials
    • Tranferring liquids and solids
    • Opening and draining pumps valves or lines
    • Loading and unloading of process equipment
    • Raw material sample collection
    • Concentration mixing and palletisation
    • Hazmat response
    • Assembly and disassembly of large pipes

Specifications

Type Protective Apron
Size 4X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Hook and Loop
Sterility Non-sterile
Cuff Style Double Inner Knit
Certifications/Compliance CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)
Weight (Metric) 103 gsm
Seam Type Ultrasonically Welded and Taped