missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-224 Two-Tone Double-Dipped Work Gloves, Vend Pack
Two-tone double-dipped work gloves, with a combination of neoprene and natural rubber.
Supplier: Ansell 87224090VEND
Description
Key Features and Benefits
- Neoprene and natural rubber blend: Enhanced chemical resistance
- 0.68 mm/27 mil thickness: Greater mechanical protection
- 100% cotton flock lining: Sweat absorption, for better comfort
Description
- Specialized protection: AlphaTec™ 87-224 industrial gloves are two-toned and double-dipped, blending neoprene and natural rubber to offer personal protection against various chemicals
- Enhanced defenses: Thanks to their mediumweight thickness of 0.68 mm/27 mil, they also provide a robust barrier against mechanical hazards
- Elevated comfort: Designed with 100% cotton flock lining, these sturdy work gloves absorb wearers' sweat, for greater comfort with sustained use
- Strengthened grip: These AlphaTec™ safety gloves also feature a honeycomb pattern finish, boosting grip on wet and dry surfaces
Specifications
| EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016 | |
| No | |
| Fully coated | |
| Neoprene | |
| Partially Textured | |
| No | |
| Mediumweight Gloves | |
| Maintenance of Plant and Heavy Equipment, Laboratory Maintenance and Clean Up, Janitorial Use, Chemical Handling | |
| Straight | |
| Cotton Flocking | |
| 87-224 | |
| 12.8 in. |
| CE 0493, EN 388:2016 +A1:2018, EN 421:2010 | |
| No | |
| Natural Rubber Latex | |
| Blue | |
| Specific | |
| Latex | |
| 9 | |
| Yellow | |
| Honeycomb | |
| Chemical, Liquid, Abrasion, Viral | |
| Keystone | |
| 325 mm |