missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-224 Two-Tone Double-Dipped Work Gloves, Vend Pack

Catalog No. 17609990
Change view
Click to view available options
Size:
10
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17609990 7
17609991 8
17609992 9
17609993 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17609990 Supplier Ansell Supplier No. 87224070VEND
Only null left

Two-tone double-dipped work gloves, with a combination of neoprene and natural rubber.

Key Features and Benefits

  • Neoprene and natural rubber blend: Enhanced chemical resistance
  • 0.68 mm/27 mil thickness: Greater mechanical protection
  • 100% cotton flock lining: Sweat absorption, for better comfort

Description

  • Specialized protection: AlphaTec™ 87-224 industrial gloves are two-toned and double-dipped, blending neoprene and natural rubber to offer personal protection against various chemicals
  • Enhanced defenses: Thanks to their mediumweight thickness of 0.68 mm/27 mil, they also provide a robust barrier against mechanical hazards
  • Elevated comfort: Designed with 100% cotton flock lining, these sturdy work gloves absorb wearers' sweat, for greater comfort with sustained use
  • Strengthened grip: These AlphaTec™ safety gloves also feature a honeycomb pattern finish, boosting grip on wet and dry surfaces

Specifications

Certifications/Compliance EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016
Certifications/Compliance (Europe) CE 0493, EN 388:2016 +A1:2018, EN 421:2010
Chemotherapeutic Rated No
Chlorinated No
Coated Area Fully coated
Cuff Material Natural Rubber Latex
Coating Material Neoprene
Color Blue
Exterior Finish Partially Textured
Hand Specific
Latex Free No
Material Latex
Product Type Mediumweight Gloves
Size 7
For Use With (Application) Maintenance of Plant and Heavy Equipment, Laboratory Maintenance and Clean Up, Janitorial Use, Chemical Handling
Glove Cuff Color Yellow
Glove Cuff Style Straight
Grip Design Honeycomb
Liner Material Cotton Flocking
Resistant To Chemical, Liquid, Abrasion, Viral
Series 87-224
Thumb Style Keystone
Length (English) 12.8 in.
Length (Metric) 325 mm
Show More Show Less