missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-085 Natural Rubber Gloves
Flocked natural rubber gloves, offering elevated comfort and food handling compliance.
$422.66 - $422.66
Specifications
| Certifications/Compliance | EN ISO 374-1:2016, EN ISO 374-5:2016, Food Type: Rubber, Category III |
|---|---|
| Certifications/Compliance (Europe) | CE 0493, EN 388:2016 +A1:2018, REACH Compliant |
| Chlorinated | Yes |
| Color | Pink |
| Finger Thickness (English) | 18 mil |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-609-981
|
Ansell
87085070BP |
7 |
Case of 144 Pair for $422.66
|
|
|||||
|
17-609-982
|
Ansell
87085080BP |
8 |
Case of 144 Pair for $422.66
|
|
|||||
|
17-609-983
|
Ansell
87085090BP |
9 |
Case of 144 Pair for $422.66
|
|
|||||
|
17-609-984
|
Ansell
87085100BP |
10 |
Case of 144 Pair for $422.66
|
|
|||||
Description
Key Features and Benefits
- Beaded cuff: Easy-to-don, tear and chemical-resistant gloves
- Food-safe: Ideal for processing food and protecting hands
- Good abrasion resistance: Designed for use in low-hazard areas
Description
- Increased protection and convenience: AlphaTec™ 87-085 rubber work gloves are flexible, lightweight and feature tear-resistant beaded cuffs for easy donning and doffing
- Food handling compliance: As these unsupported protective gloves possess an AQL of 1.5, they are also suitable for food processing applications
- Added durability: AlphaTec™ 87-085 abrasion-resistant gloves offer moderate durability when working in agriculture, food processing and chemical industries
- Improved comfort: These pink safety gloves feature 100% cotton flock lining, which ensures flexibility and elevated comfort
Specifications
| EN ISO 374-1:2016, EN ISO 374-5:2016, Food Type: Rubber, Category III | |
| Yes | |
| 18 mil | |
| No | |
| 18 mil | |
| Chemical Protection Gloves | |
| 0.45 mm | |
| No | |
| KLT | |
| Beaded | |
| Unlined | |
| 300 mm |
| CE 0493, EN 388:2016 +A1:2018, REACH Compliant | |
| Pink | |
| 0.45 mm | |
| Rubber Latex | |
| 0.45 mm | |
| 18 mil | |
| EN 388:2016 +A1:2018 - 2010X | |
| Type A | |
| Bacteria, Fungi only | |
| Raised Diamond | |
| 11.8 in. |