Learn More
Ansell AlphaTec™ 58-435 Heavy Duty Nitrile Gloves
Catalog No. 17999819
Change view
Click to view available options
Size:
10
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999819 10
17999816 7
17999817 8
17999818 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17999819 Supplier Ansell Supplier No. 58435100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Heavy-duty nitrile gloves with ANSELL GRIP™ technology features a 16 mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters.
  • ANSELL GRIP™ Technology for optimal control when handling oily or wet objects
  • Premium grade nitrile shell offers excellent chemical protection against a wide range of chemicals
  • Lightweight, with cotton flocked lining offering superior comfort
  • Enhanced grip, dexterity and comfort helps reduce hand fatigue
  • Extra-long cuff extends protection to the forearm
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III, EN 1149
Chlorinated No
Color Gray
Cut Resistance Rating ANSI A5
Latex Free Yes
Material Nitrile
Size 10
EN388 Standard EN 388
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design ANSELL GRIP™ Technology
Liner Material Cotton Flock
Lining Lined
Model 256251
Series AlphaTec™ 58-435
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less