missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-201 Green Nitrile Gloves
GREENER_CHOICE

Catalog No. 17033226
Change view
Click to view available options
Size:
10
8
9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17033226 10
17033224 8
17033225 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17033226 Supplier Ansell Supplier No. 58201100

Utilizing a unique formulation, AlphaTec 58-201 is a truly versatile protective solution that can be applied to a wide range of applications and environments.

  • AnsellGrip Technology for enhanced grip in Wet and Oily conditions.
  • Designed to protect at temperatures from -40 to +250°C/482°F.
  • Acrylic liner keeps the hands warm in cold conditions and protected in contact heat applications.
  • Nitrile chemical barrier providing protection against a wide range of chemicals.

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance ANSI ABR 5, ANSI Cut A2, CE 0493, EN 388:2016 +A1:2018, EN 374:2003, EN 388:2003, EN407:2020 , EN 511:2006, Food Contact
Applications Industrial
Color Green
Latex Free Yes
Material Acrylic, Nitrile
Product Type Chemical-resistant Gloves
Size 10
Glove Cuff Style Gauntlet
Grip Design Ansell Grip Technology
Liner Material Acrylic
Length (Metric) 330 mm
Show More Show Less