missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ ALPHATEC™ 53-002
Description
- Unique rubber polymer and plastic laminate technology delivering maximum chemical protection against an extremely broad set of chemical types.
- Unique manufacturing process allows for the glove to be form fitting.
- Raised diamond palm pattern for enhanced grip.
- Unique donning and doffing layer to allow ease of use.
- Anti-static according to EN 1149-3.
- Natural rubber and silicone free.
Specifications
Specifications
| Certifications/Compliance | CE 0493, EN ISO 374-5:2016, EN 388:2016 +A1:2018, EN 421:2010, EN ISO 374-1:2016 |
| Chlorinated | No |
| Description | AlphaTec 53-002 the revolutionary glove that redefines chemical protection. Our innovative design combines an advanced rubber polymer coating with cutting-edge plastic laminate technology, setting a new benchmark in chemical permeation performance whilst delivering comfort and tactility. |
| Applications | Laboratory |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Product Type | Neoprene Gloves |
| Size | 9 |
| Thickness (English) | 17 mil |
| Show More |