missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 53-001 Nitrile Chemical Resistant Gloves
Black gloves feature an innovative multi-layer polymer design to offer multi-barrier chemical protection.
Supplier: Ansell 53001080NA
Description
- Innovative multi-layer polymer design - nitrile/neoprene/nitrile layers
- Provides chemical protection against a wide range of chemicals from acids and bases to hydrocarbons and organic solvent
- Tested according to EN ISO 374:2016, permeation time > 30 minutes against 13 chemicals out of 18 listed
- Features the latest MICROCHEM™ chemical barrier technology providing superior protection for use in hazardous environments
- ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
- Inner soft nylon liner for comfort and increased mechanical protection
- Oeko-Tex™ Standard 100 Approved
Specifications
| ANSI 3 | |
| No | |
| Yes | |
| 8 | |
| 0.38 mm | |
| No | |
| ACFGJKLMNOPST | |
| Gauntlet | |
| Nylon | |
| 53001080-NA | |
| 13 in. |
| EN 420:2003 + A1:2009, EN 1149, EN407 | |
| Gray, Black | |
| Nitrile, Neoprene | |
| 15 mil | |
| EN 388:3121A | |
| Type A | |
| Bacteria, Fungi only | |
| Ansell Grip Technology | |
| Lined | |
| AlphaTec™ 53-001 | |
| 35 cm |