missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 53-001 Nitrile Chemical Resistant Gloves

Catalog No. 19500933
Change view
Click to view available options
Size:
10
11
7
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19500933 7
19500934 8
19500935 9
19500936 10
19500937 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19500933 Supplier Ansell Supplier No. 53001070NA

Black gloves feature an innovative multi-layer polymer design to offer multi-barrier chemical protection.

  • Innovative multi-layer polymer design - nitrile/neoprene/nitrile layers
  • Provides chemical protection against a wide range of chemicals from acids and bases to hydrocarbons and organic solvent
  • Tested according to EN ISO 374:2016, permeation time > 30 minutes against 13 chemicals out of 18 listed
  • Features the latest MICROCHEM™ chemical barrier technology providing superior protection for use in hazardous environments
  • ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
  • Inner soft nylon liner for comfort and increased mechanical protection
  • Oeko-Tex™ Standard 100 Approved
Note:  Size equivalents are as follows: 7 (S), 8 (M), 9 (L), 10 (XL), 11 (2XL).

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 1149, EN407
Chlorinated No
Color Gray, Black
Latex Free Yes
Material Nitrile, Neoprene
Size 7
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:3121A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ACFGJKLMNOPST
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Ansell Grip Technology
Liner Material Nylon
Lining Lined
Model 53001070-NA
Series AlphaTec™ 53-001
Length (English) 13 in.
Length (Metric) 35 cm
Show More Show Less