missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 4000 CFR Stitched and Taped Suit

Catalog No. 17033265
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17033265 5X-Large
17033258 Small
17033259 Medium
17033260 Large
17033261 X-Large
17033262 2X-Large
17033263 3X-Large
17033264 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17033265 Supplier Ansell Supplier No. OR47T9211109
Only null left

AlphaTec™ 4000 CFR Stitched and Taped - Model 111 Comfortable, anti-static chemical protective suit with exceptional multi-hazard solutions.

  • Multi-hazard solution: Excellent chemical protection
  • Flame-retardant: Protected from chemical spray by limiting flame spread
  • Optimized body fit: Improved wearer comfort and safety

Specifications

Product Type FR/CP Coveralls
Size 5X-Large
Gender Unisex
Material FR SMMS
Color Fluorescent Orange
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Double Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals
Coating Uncoated
Certifications/Compliance 16 CFR1610, AATCC76, ASTM D6413, ASTM F1671, CE 0598, EN ISO 13688:2013, EN ISO 14116:2015 Index 1
Description Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8-hour barrier against chemicals such as non-flammable, flammable and aggressive solvents.
Latex Free Yes
No. of Pockets None
For Use With (Application) Hazmat response, Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of process equipment, Raw material sample collection, Concentration mixing and palletisation, Assembly and disassembly of large pipes
Height (English) 78 in.
Height (Metric) 198.12 cm
Weight (English) 320 lb.
Packaging Individually Packed
Style Protective Suits
Show More Show Less