missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 4000 CFR Stitched and Taped Suit

AlphaTec™ 4000 CFR Stitched and Taped - Model 111 Comfortable, anti-static chemical protective suit with exceptional multi-hazard solutions.

Supplier:  Ansell OR47T9211108

 View more versions of this product

Catalog No. 17-033-264

Only null left
Explore available promotions

Description

Description

  • Multi-hazard solution: Excellent chemical protection
  • Flame-retardant: Protected from chemical spray by limiting flame spread
  • Optimized body fit: Improved wearer comfort and safety
Specifications

Specifications

FR/CP Coveralls
Unisex
Fluorescent Orange
Elastic
Elastic
Elastic
Stitched
Chemicals
16 CFR1610, AATCC76, ASTM D6413, ASTM F1671, CE 0598, EN ISO 13688:2013, EN ISO 14116:2015 Index 1
Yes
Hazmat response, Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of process equipment, Raw material sample collection, Concentration mixing and palletisation, Assembly and disassembly of large pipes
192.02 cm
Individually Packed
4X-Large
FR SMMS
Limited-use
Elastic
Hooded
Double Zipper
Non-sterile
Uncoated
Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8-hour barrier against chemicals such as non-flammable, flammable and aggressive solvents.
None
75.6 in
300 lb.
Protective Suits
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions