missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 CFR Stitched and Taped Suit
AlphaTec™ 4000 CFR Stitched and Taped - Model 111 Comfortable, anti-static chemical protective suit with exceptional multi-hazard solutions.
Supplier: Ansell OR47T9211106
Description
- Multi-hazard solution: Excellent chemical protection
- Flame-retardant: Protected from chemical spray by limiting flame spread
- Optimized body fit: Improved wearer comfort and safety
Specifications
| FR/CP Coveralls | |
| Unisex | |
| Fluorescent Orange | |
| Elastic | |
| Elastic | |
| Elastic | |
| Stitched | |
| Chemicals | |
| 16 CFR1610, AATCC76, ASTM D6413, ASTM F1671, CE 0598, EN ISO 13688:2013, EN ISO 14116:2015 Index 1 | |
| Yes | |
| Hazmat response, Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of process equipment, Raw material sample collection, Concentration mixing and palletisation, Assembly and disassembly of large pipes | |
| 182.8 cm | |
| Individually Packed |
| 2X-Large | |
| FR SMMS | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Double Zipper | |
| Non-sterile | |
| Uncoated | |
| Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8-hour barrier against chemicals such as non-flammable, flammable and aggressive solvents. | |
| None | |
| 72 in. | |
| 240 lb. | |
| Protective Suits |