missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 CFR Stitched and Taped Suit
AlphaTec™ 4000 CFR Stitched and Taped - Model 111 Comfortable, anti-static chemical protective suit with exceptional multi-hazard solutions.
Supplier: Ansell OR47T9211103
Description
- Multi-hazard solution: Excellent chemical protection
- Flame-retardant: Protected from chemical spray by limiting flame spread
- Optimized body fit: Improved wearer comfort and safety
Specifications
| FR/CP Coveralls | |
| Unisex | |
| Fluorescent Orange | |
| Elastic | |
| Elastic | |
| Elastic | |
| Stitched, Taped | |
| Chemicals | |
| 16 CFR1610, AATCC76, ASTM D6413, ASTM F1671, CE 0598, EN ISO 13688:2013, EN ISO 14116:2015 Index 1 | |
| Yes | |
| Elevated comfort: The optimized body fit design of this protective suit offers improved wearer comfort and safety. This full-chemical protection suit defends against infective agents, tested according to ASTM F1671 and hydrocarbons such as hexane, HF, oleum and many more. | |
| 158.5 cm | |
| Individually Packed |
| Medium | |
| FR SMMS | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Double Zipper | |
| Non-sterile | |
| Uncoated | |
| Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8-hour barrier against chemicals such as non-flammable, flammable and aggressive solvents. | |
| None | |
| 62.4 in. | |
| 160 lb. | |
| Protective Suits |