missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 39-122 Series Nitrile Heavy Duty Gloves

Catalog No. p-9012025
Change view
Click to view available options
Size:
9
10
Catalog No. Size
1139438C 9
1139438D 10
2 options

Catalog No. 1139438C

Supplier: Ansell 3912211090

Only null left

Provides heavy-duty mechanical and chemical protection for wide variety of work environments. Sol-Knit™ Nitrile Heavy Duty Gloves have better mechanical performance and chemical protection.

  • Liquid and chemical protection gloves
  • Industrial gloves
  • Tough glove offers optimum comfort of use thanks to its natural shape that makes it well suited for use with long-lasting tasks
  • Superior dexterity and the high levels of wet and dry grip contribute to wearer safety and ultimately to the overall productivity
  • Rugged glove with chemical protection

Applications:

  • Adjusting panels parts components
  • Applying finish to materials, products
  • Assembly and disassembly of equipment
  • Equipment repair, maintenance
  • Handling materials with sharp or rough edges
  • Oil, fluids and filter change
  • Production line support and maintenance

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Chlorinated No
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 9
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Lining Lined
Model 39-122 9 217801
Series AlphaTec™ 39-122
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less