missing translation for 'onlineSavingsMsg'
Learn More

Ansell ALPHATEC™ 1800 COMFORT Bound - Model 195 Coverall White/Navy

Catalog No. 296369286
Change view
Click to view available options
Size:
Medium
Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296369286 2X-Large
296369281 Medium
296369283 Large
296369282 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296369286 Supplier Ansell Supplier No. WN18B9219506
Only null left

Lightweight, breathable protective body suit, offering wearers light chemical resistance

  • Particle barrier: Resists low-hazard liquid spray and fine particulates
  • Microporous laminate with SMS features: For heat stress protection
  • Bound seams: Heightened strength and barrier protection

Specifications

Product Type Controlled Environments ESD Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Non-woven Polypropylene and Polyethylene Laminate
Color White
Disposable Reusable
Ankle Style Bound
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance |EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|AATCC76|CE Number|EN 1073-2:2002|EN 1149-5:2008|US Ergo Certification
Latex Free Yes
No. of Pockets None
For Use With (Application) Grants protection against fine particulates and low-hazard liquid spray
Length (English) 5 in.
Length (Metric) 12.7 cm
Height (English) 4 in.
Height (Metric) 10.16 cm
Show More Show Less