missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell ALPHATEC™ 1800 COMFORT Bound - Model 195 Coverall White/Navy
Lightweight, breathable protective body suit, offering wearers light chemical resistance
Supplier: Ansell WN18B9219504
Description
- Particle barrier: Resists low-hazard liquid spray and fine particulates
- Microporous laminate with SMS features: For heat stress protection
- Bound seams: Heightened strength and barrier protection
Specifications
| Controlled Environments ESD Coveralls | |
| Unisex | |
| White | |
| Bound | |
| Hooded | |
| Bound | |
| Chemicals | |
| Yes | |
| Grants protection against fine particulates and low-hazard liquid spray | |
| 12.7 cm | |
| 10.16 cm |
| Large | |
| Microporous Non-woven Polypropylene and Polyethylene Laminate | |
| Reusable | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| |EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|AATCC76|CE Number|EN 1073-2:2002|EN 1149-5:2008|US Ergo Certification | |
| None | |
| 5 in. | |
| 4 in. |