missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 1500 PLUS Stitched Model 111, Coveralls with Hood and Finger Loops

Catalog No. p-200003556
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
17010440 4X-Large
17010441 Small
17010442 Large
17010443 5X-Large
17010444 Medium
17010445 X-Large
17010446 2X-Large
17010447 3X-Large
8 options

Catalog No. 17010440

Supplier: Ansell WH15S9211108

Only null left

Particle-filtering, anti-static SMS protective suit delivers light chemical resistance.

  • Particle-filtering SMS fabric: Filters 100% of particles over 3 microns(According to KAKEN particle penetration test)
  • Air and water vapor permeability: Ensures heat stress protection
  • Optimized body fit: Enhanced comfort

Specifications

Product Type Single-use Coveralls
Size 4X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Particle, Anti-static
Certifications/Compliance (Europe) CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
Latex Free No
Model WH15-S-92-111
No. of Pockets None
For Use With (Application) Machinery and Equipment, Life Sciences, Metal Fabrication, Construction, Chemical
Show More Show Less