Learn More
AlphaProTech Critical Cover™ Microbreathe™ SafeStep™ Boot Covers
Catalog No. p-4909806
Change view
Click to view available options
Size:
Large
X-Large
Thread Color:
Black
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Color
19132211 Large Orange
19132213 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19132211 Supplier AlphaProTech Supplier No. BT A1616 B
Only null left
Anti-skid boot covers fit over the calf with an elastic top to provide a comfortable fit and to prevent the boot cover from sliding down.
  • Proprietary high-performing, environmentally friendly boot cover
  • Inherent non-skid physical properties
  • Strong enough to withstand even the most abrasive floor surfaces
  • Fabric provides barrier protection and exhibits low particle generation
  • Four sizes to choose from to ensure a proper fit
  • Different thread colors around the opening for convenient size identification
Specifications
Size Large
Material Polyethylene, Polypropylene
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Anti-skid
Seam Type Serged
Thread Color Orange
Packaging Quantity 100 Pairs/Cs.
Length (English) 12.25 in.
Length (Metric) 31.1 cm
Height (English) 17.5 in.
Height (Metric) 44.45 cm
Show More Show Less