missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ GenPro™ Frocks

Catalog No. 191301516F
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Length (Metric) Sleeve:
87.63 cm
90.17 cm
92.71 cm
95.25 cm
101.6 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (Metric) Sleeve
191301516F 3X-Large 101.6 cm
191301516B Medium 87.63 cm
191301516C Large 90.17 cm
191301516D X-Large 92.71 cm
191301516E 2X-Large 95.25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 191301516F Supplier AlphaProTech Supplier No. FK121226

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polypropylene frocks offers protection for non-hazardous environments. Ideal for facility visitors.

  • Polypropylene frock ideal for personnel or facility visitors
  • Offer comfort and breathability
  • Extra long design provides additional coverage
  • No pockets to help control contaminants from entering or leaving sensitive areas
  • Elastic wrist
  • Snap mandarin collar
  • Five-snap closure

Specifications

Size 3X-Large
Material Composite Fabric
Color White
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Snap Mandarin
Size (English) Chest 29 in.
Size (Metric) Chest 73.6 cm
Sleeve Style Raglan Sleeve
Length (Metric) Sleeve 101.6 cm
Product Type Controlled Environments Frock
Seam Type Serged
Show More Show Less