missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ GenPro™ Bouffants

Catalog No. p-3741852
Change view
Click to view available options
Size:
21 in.
24 in.
26 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191301507 21 in.
191301510 24 in.
191304188 26 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191301507 Supplier AlphaProTech Supplier No. BO 13821 B
Only null left

Bouffants available in three different sizes are made of soft polypropylene material and provide the appropriate level of protection required to contain particles resulting from the head area.

  • Basic protection for non-hazardous environments
  • Non-binding, latex-free elastic
  • Secure fit
  • Serged seams
  • Lightweight material
  • White

Specifications

Product Type Cleanroom Bouffant
Size 21 in.
Material Polypropylene
Color White
Disposable Single-use
Design Type Non-binding elastic opening
Latex Free Yes
Model BO-138
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance 21C CFR Part 820
Packaging Type Bulk
Quantity 1000/Cs.
Includes Non-binding, Latex-free Band
Construction Round
For Use With (Application) Dry, Non-hazardous Environments
Resistance Elastic
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less