missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Beard Cover
Description
- For non-hazardous environments
- Strong, lightweight and breathable
- Latex-free
- Limited barrier to particulates and contamination
Specifications
Specifications
| Size | Universal |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Latex Free | Yes |
| Seam Type | Universal |
| Certifications/Compliance | 21C CFR Part 820 |
| For Use With (Application) | Dry, Non-hazardous Environments |
| Quantity | 1000 |
| Product Type | Cleanroom Beard Cover |