missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ ComforTech™ Sleeves

Catalog No. 296358526
Change view
Click to view available options
Size:
Universal
X-Large
Cuff Style:
Elastic
Elastic with Thumb Loop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style
296358526 Universal Elastic
191304267 X-Large Elastic
191694054 Universal Elastic with Thumb Loop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296358526 Supplier AlphaProTech Supplier No. MS01J068

Protective sleeves are constructed of a high quality microporous composite material with a low particle count to meet the requirements of controlled environments.

  • Low particle count
  • Tapered sleeve with elastic at both ends
  • Sonic welded seams
  • ComforTech material is made of microporous material for use in cleanrooms and controlled environments

Specifications

Product Type Sleeve Protector
Size Universal
Construction Tapered
Material Microporous Laminate
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Ultrasonically Welded
Cuff Style Elastic
Latex Free Yes
Model MS-01J06
For Use With (Application) Cleanrooms/Controlled Environments
Certifications/Compliance 21C CFR Part 820
Length (English) 19.5 in.
Length (Metric) 49.5 cm
Packaging Bulk
No. per Case 300
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less