Learn More
AlphaProTech Critical Cover™ ComforTech™ Lab Coats
Strong, lightweight and breathable ComforTech material provides protection from fluids, light chemical splash, particles and biologicals without compromising comfort.
Supplier:  AlphaProTech LCJ26213
 View more versions of this product
Catalog No. 19-168-171C
Only null left
Explore available promotions
Description
Description
  • Strong, lightweight and breathable ComforTech material
  • Protection from fluids, light chemical splash, particles and biologicals
  • Low particle count and high filtration efficiency of 99.9%
  • Features serged seams, five-snap closure, elastic wrists, tapered collar, and three pockets
  • ComforTech is composed of breathable polyethylene film, spunbond polypropylene nonwoven laminated
Specifications
Specifications
Controlled Environments Lab Coat
Single-use
Fluids, Light Chemical Splash, Particulates, Biologicals
Microporous Composite Fabric
Non-sterile
40 in.
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'