missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ ComforTech™ Lab Coats
Strong, lightweight and breathable ComforTech material provides protection from fluids, light chemical splash, particles and biologicals without compromising comfort.
Supplier: AlphaProTech LCJ26212
Description
- Strong, lightweight and breathable ComforTech material
- Protection from fluids, light chemical splash, particles and biologicals
- Low particle count and high filtration efficiency of 99.9%
- Features serged seams, five-snap closure, elastic wrists, tapered collar, and three pockets
- ComforTech is composed of breathable polyethylene film, spunbond polypropylene nonwoven laminated
Specifications
| Controlled Environments Lab Coat | |
| Single-use | |
| Fluids, Light Chemical Splash, Particulates, Biologicals | |
| 100.3 cm |
| Microporous Composite Fabric | |
| Non-sterile | |
| 39.5 in. |