missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ BarrierTech™ MaxGrip™ Shoe Covers

Catalog No. p-3879528
Change view
Click to view available options
Size:
Universal
Small
Medium
Large
X-Large
Thread Color:
Black
Green
Orange
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Color
19811001 Universal White
19811000 Small Green
19811003 Medium Yellow
19811004 Large Orange
19811002 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19811001 Supplier AlphaProTech Supplier No. SHE1W12BH
Only null left

Effectively prevent contaminants from entering your environment whlie providing secure footing with MaxGrip™ skid-resistant sole. Available in five different sizes.

  • Serged seams
  • Anti-skid
  • Impervious material
  • Thread color for easy identification

Specifications

Size Universal
Material BarrierTech™
Color White
Disposable Single-use
Sterility Non-sterile
Sole Type Anti-skid
Seam Type Serged
Thread Color White
Packaging Quantity 100 Pairs/Cs.
Dimensions (L x W x H) 14.25 x 4.5 x 4.75 in. (36.19 x 11.43 x 12.06cm)
Length (English) 14.25 in.
Length (Metric) 36.19 cm
Height (English) 4.75 in.
Height (Metric) 12.06 cm
Product Type Controlled Environments Shoe Covers
Show More Show Less