missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Hood
Description
- Lightweight, breathable material
- Comfortable, nonbinding latex-free elastic bands around face and neck for secure fit
- Provide secure complete coverage and comfort without pinching or binding
- Open face style provides complete coverage without obstructing vision or range of motion
Specifications
Specifications
| Product Type | Combination Head and Beard Cover |
| Size | X-Large |
| Material | Polypropylene |
| Color | Blue |
| Design Type | Elastic |
| Latex Free | Yes |
| Seam Type | Serged |
| Sterility | Non-sterile |
| Certifications/Compliance | 21C CFR Part 820 |
| Quantity | 500/Cs. |
| Show More |