Learn More
AlphaProTech Critical Cover™ GenPro™ Hood
Catalog No. 19810375
Change view
Click to view available options
Color:
Blue
White
Size:
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19810375 Blue X-Large
19810370 White X-Large
19140230 White 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19810375 Supplier AlphaProTech Supplier No. SD 92342 B
Designed to protect against dirt, grime, and particulates in nonhazardous environments.
  • Lightweight, breathable material
  • Comfortable, nonbinding latex-free elastic bands around face and neck for secure fit
  • Provide secure complete coverage and comfort without pinching or binding
  • Open face style provides complete coverage without obstructing vision or range of motion
Specifications
Product Type Combination Head and Beard Cover
Size X-Large
Material Polypropylene
Color Blue
Design Type Elastic
Latex Free Yes
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance 21C CFR Part 820
Quantity 500/Cs.
Style Bouffant and Beard Cover
Show More Show Less